Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neighbor of BRCA1 gene 2 (non-protein coding) | NBR2 | NCRNA00192 | Sense | Chr. 17 - 43125583 - 43145113 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101929767 | LOC101929767 | 10.0kb | Anti-sense | Chr. 17 - 43148366 - 43170403 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map