Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | trafficking protein particle complex 9 | TRAPPC9 |
IBP, IKBKBBP, MRT13, NIBP, T1, TRS120 |
Anti-sense | Chr. 8 - 139727725 - 140458579 on NCBI Build GRCh38 | |
| Downstream | chromatin accessibility complex 1 | CHRAC1 |
CHARC1, CHARC15, CHRAC-1, CHRAC-15, CHRAC15, YCL1 |
415.8kb | Sense | Chr. 8 - 140511298 - 140517153 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map