Assay Details
| Gene Symbol: ADAM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF334867.1 | - | 1 | 418 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927228 | LOC101927228 | -47.8kb | Anti-sense | Chr. 16 - 49466106 - 49469875 on NCBI Build GRCh38 | |
| Downstream | CTD nuclear envelope phosphatase 1 regulatory subunit 1 | CNEP1R1 |
C16orf69, NEP1-R1, NEP1R1, TMEM188, TMP125 |
506.6kb | Anti-sense | Chr. 16 - 50025206 - 50037088 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

ADAM3B
Genomic Map