Assay Details
| Gene Symbol: LINC00519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BG121773.1 | - | 1 - 2 | 262 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thioredoxin related transmembrane protein 1 | TMX1 |
PDIA11, TMX, TXNDC, TXNDC1 |
-46.7kb | Anti-sense | Chr. 14 - 51240168 - 51257655 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 640 | LINC00640 | 5.1kb | Anti-sense | Chr. 14 - 51333393 - 51365557 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC00519
Genomic Map