Assay Details
| Gene Symbol: LSINCT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | GU228577.2 | - | 1 | 1819 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100506858 | LOC100506858 | -400.4kb | Anti-sense | Chr. 5 - 2303720 - 2312201 on NCBI Build GRCh38 | |
| Downstream | chromosome 5 open reading frame 38 | C5orf38 |
CEI, IRX2NB |
36.7kb | Sense | Chr. 5 - 2751940 - 2758576 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LSINCT5
Genomic Map