Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin conjugating enzyme E2 S | UBE2S |
E2-EPF, E2EPF, EPF5 |
Sense | Chr. 19 - 55401282 - 55408146 on NCBI Build GRCh38 | |
| Downstream | shisa family member 7 | SHISA7 | 25.7kb | Sense | Chr. 19 - 55428738 - 55442863 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map