Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphodiesterase 4D interacting protein pseudogene | LOC728989 | Sense | Chr. 1 - 147019336 - 147043039 on NCBI Build GRCh38 | ||
| Downstream |
RNA, variant U1 small nuclear 8 |
RNVU1-8 |
RNU1-145, vU1.8 |
63.2kb | Sense | Chr. 1 - 147084616 - 147084756 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map