Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 302c | MIR302C |
MIRN302C, mir-302c |
-83.0 | Sense | Chr. 4 - 112648363 - 112648430 on NCBI Build GRCh38 |
| Downstream |
ankyrin 2, neuronal |
ANK2 |
ANK-2, LQT4, brank-2 |
56.0kb | Anti-sense | Chr. 4 - 112706083 - 113383740 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map