Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 367 | MIR367 |
MIRN367, hsa-mir-367 |
Sense | Chr. 4 - 112647874 - 112647941 on NCBI Build GRCh38 | |
| Downstream |
ankyrin 2, neuronal |
ANK2 |
ANK-2, LQT4, brank-2 |
57.4kb | Anti-sense | Chr. 4 - 112706083 - 113383740 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map