Assay Details
| Gene Symbol: SMARCA5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104027.1 | 1 - 2 | 394 | 115 | ||
| GenBank mRNA | AK307172.1 | - | 1 - 2 | 164 | 115 | |
| HY014185.1 | - | 1 - 2 | 394 | 115 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5 |
SMARCA5 |
ISWI, SNF2H, WCRF135, hISWI, hSNF2H |
Anti-sense | Chr. 4 - 143513463 - 143557489 on NCBI Build GRCh38 | |
| Downstream |
glucuronidase, beta pseudogene 5 |
GUSBP5 | GYPELOC441046 | 44.6kb | Anti-sense | Chr. 4 - 143559472 - 143561460 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012009.hg.1 | ENST00000500800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012009.hg.1 | ENST00000500800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012009.hg.1 | NR_104027.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012009.hg.1 | NR_104027.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012009.hg.1 | NR_104027.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012009.hg.1 | ENST00000500800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012009.hg.1 | ENST00000500800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012009.hg.1 | NR_104027.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012009.hg.1 | NR_104027.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012009.hg.1 | NR_104027.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |

SMARCA5-AS1
Genomic Map