Assay Details
| Gene Symbol: SNRNP35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104103.1 | 1 - 2 | 110 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rab interacting lysosomal protein like 2 | RILPL2 | RLP2 | -21.4kb | Anti-sense | Chr. 12 - 123408567 - 123436733 on NCBI Build GRCh38 |
| Downstream | microRNA 3908 | MIR3908 | 64.0kb | Sense | Chr. 12 - 123536409 - 123536534 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000350887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000350887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000350887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000412157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000412157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000412157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000412157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000412157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000527158.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000527158.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000527158.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000527158.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000529201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000529201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000529201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000529201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000529201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000529904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000529904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000529904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000529904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | ENST00000529904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | NM_022717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | NM_022717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | NM_022717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | NM_022717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | NM_180699.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | NM_180699.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | NM_180699.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | NM_180699.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | NR_104103.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | NR_104103.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009242.hg.1 | NR_104103.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000544468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000544468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000544468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000544468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000544468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000610631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000623184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000623184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | ENST00000623184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007436.hg.1 | ENST00000584258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007436.hg.1 | ENST00000584258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000350887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000350887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000350887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000412157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000412157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000412157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000412157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000412157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000526639.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000527158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000527158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000527158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000527158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000529201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000529201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000529201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000529201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000529201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000529904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000529904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000529904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000529904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | ENST00000529904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | NM_022717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | NM_022717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | NM_022717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | NM_022717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | NM_180699.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | NM_180699.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | NM_180699.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | NM_180699.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | NR_104103.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | NR_104103.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009242.hg.1 | NR_104103.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000340724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000376874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000544468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000544468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000544468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000544468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000544468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000610631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000623184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000623184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | ENST00000623184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012273.hg.1 | NM_178314.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007436.hg.1 | ENST00000584258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007436.hg.1 | ENST00000584258.1 | Ensembl |

SNRNP35
Genomic Map