Assay Details
| Gene Symbol: LOC101927190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104675.1 | 1 - 2 | 91 | 63 | ||
| GenBank mRNA | BX649078.1 | - | 1 - 2 | 92 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADP ribosylation factor like GTPase 14 effector protein like | ARL14EPL | -313.0kb | Anti-sense | Chr. 5 - 116051140 - 116060023 on NCBI Build GRCh38 | |
| Downstream | semaphorin 6A | SEMA6A |
HT018, SEMA, SEMA6A1, SEMAQ, VIA |
69.0kb | Sense | Chr. 5 - 116443555 - 116574926 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927190
Genomic Map