Assay Details
Gene Symbol: PRDX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126103.1 | 1 - 2 | 107 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sideroflexin 4 | SFXN4 |
BCRM1, COXPD18 |
-8.0kb | Sense | Chr. 10 - 119140913 - 119165711 on NCBI Build GRCh38 |
Downstream | G protein-coupled receptor kinase 5 | GRK5 | GPRK5 | 28.8kb | Anti-sense | Chr. 10 - 119207685 - 119455619 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
maternal placenta development
apoptotic process
response to oxidative stress
mitochondrion organization
positive regulation of cell proliferation
peptidyl-cysteine oxidation
myeloid cell differentiation
response to lipopolysaccharide
negative regulation of kinase activity
cellular response to oxidative stress
cellular response to reactive oxygen species
response to hydrogen peroxide
hydrogen peroxide catabolic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
cell redox homeostasis
positive regulation of NF-kappaB transcription factor activity
regulation of mitochondrial membrane potential
cellular oxidant detoxification
protein binding
protein C-terminus binding
thioredoxin peroxidase activity
alkyl hydroperoxide reductase activity
kinase binding
protein kinase binding
identical protein binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
response to reactive oxygen species
maternal placenta development
apoptotic process
response to oxidative stress
mitochondrion organization
positive regulation of cell proliferation
peptidyl-cysteine oxidation
myeloid cell differentiation
response to lipopolysaccharide
negative regulation of kinase activity
cellular response to oxidative stress
cellular response to reactive oxygen species
response to hydrogen peroxide
hydrogen peroxide catabolic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
cell redox homeostasis
positive regulation of NF-kappaB transcription factor activity
regulation of mitochondrial membrane potential
cellular oxidant detoxification
protein binding
protein C-terminus binding
thioredoxin peroxidase activity
alkyl hydroperoxide reductase activity
kinase binding
protein kinase binding
identical protein binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000463322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000463322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000463322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000463322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000494433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000494433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000494433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000494433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | ENST00000494433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126106.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126106.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126106.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126106.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126106.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011993.hg.1 | NR_126106.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000298510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000463322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000463322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000463322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000463322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000494433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000494433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000494433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000494433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | ENST00000494433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_001302272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NM_006793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126102.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126105.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126106.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126106.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126106.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126106.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126106.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011993.hg.1 | NR_126106.1 | Refseq |
