| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000568070.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000568070.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
ENST00000568070.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000563357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000568070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000568070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
ENST00000568070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008916.hg.1 |
NR_126161.1 |
Refseq |