| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000550664.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000550664.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000550664.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000550664.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000550664.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000547418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000549568.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000550664.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000550664.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000550664.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000550664.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000550664.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
ENST00000552759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008653.hg.1 |
NR_126333.1 |
Refseq |