Assay Details
| Gene Symbol: LOC101929058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126333.1 | 5 - 6 | 584 | 109 | ||
| GenBank mRNA | AI143882.1 | - | 2 - 3 | 109 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 12 open reading frame 42 | C12orf42 | Anti-sense | Chr. 12 - 103047669 - 103525258 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC105369945 | LOC105369945 | 331.3kb | Sense | Chr. 12 - 103496040 - 103497695 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929058
Genomic Map