Assay Details
| Gene Symbol: LINC00363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126360.1 | 2 - 3 | 103 | 170 | ||
| GenBank mRNA | AA885151.1 | - | 2 - 3 | 170 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glypican 5 | GPC5 | -179.8kb | Sense | Chr. 13 - 91398619 - 92867237 on NCBI Build GRCh38 | |
| Downstream | glypican 6 | GPC6 | OMIMD1 | 170.1kb | Sense | Chr. 13 - 93226825 - 94408020 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC00363
Genomic Map