Assay Details
| Gene Symbol: LINC01309 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126374.1 | 1 - 2 | 125 | 80 | ||
| GenBank mRNA | AA421210.1 | - | 1 - 2 | 127 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 10 member 2 | SLC10A2 |
ASBT, IBAT, ISBT, NTCP2, PBAM |
-358.4kb | Anti-sense | Chr. 13 - 103043998 - 103066846 on NCBI Build GRCh38 |
| Downstream | DAOA antisense RNA 1 | DAOA-AS1 |
DAOA-AS, DAOAAS, G30 |
2.0mb | Anti-sense | Chr. 13 - 105459055 - 105505681 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01309
Genomic Map