Assay Details
| Gene Symbol: WWC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126473.1 | 1 - 2 | 104 | 101 | ||
| GenBank mRNA | BX537950.1 | - | 1 - 2 | 105 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WWC2 antisense RNA 2 | WWC2-AS2 | C4orf38 | -134.4kb | Sense | Chr. 4 - 183097021 - 183099199 on NCBI Build GRCh38 |
| Downstream | claudin 22 | CLDN22 | CLDN21 | 77.4kb | Sense | Chr. 4 - 183318067 - 183320774 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

WWC2-AS1
Genomic Map