Assay Details
| Gene Symbol: RAI1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130894.1 | 1 - 2 | 485 | 77 | ||
| GenBank mRNA | BX366317.2 | - | 1 - 2 | 492 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
Smith-Magenis syndrome chromosome region, candidate 2 (non-protein coding) |
SMCR2 | TCONS_00025215 | -81.5kb | Sense | Chr. 17 - 17674026 - 17677688 on NCBI Build GRCh38 |
| Downstream |
Smith-Magenis syndrome chromosome region, candidate 5 (non-protein coding) |
SMCR5 | NCRNA00034 | 10.4kb | Sense | Chr. 17 - 17776686 - 17779529 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

RAI1-AS1
Genomic Map