Assay Details
| Gene Symbol: SNORA88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132770.1 | 1 | 107 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | reticulocalbin 1 | RCN1 |
HEL-S-84, PIG20, RCAL, RCN |
-38.0kb | Anti-sense | Chr. 11 - 32090931 - 32105726 on NCBI Build GRCh38 |
| Downstream | Wilms tumor 1 | WT1 |
AWT1, EWS-WT1, GUD, NPHS4, WAGR, WIT-2, WT33 |
243.9kb | Sense | Chr. 11 - 32387775 - 32435535 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SNORA88
Genomic Map