Assay Details
| Gene Symbol: SNORA104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132784.1 | 1 | 85 | 110 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adhesion G protein-coupled receptor E3 | ADGRE3 | EMR3 | Anti-sense | Chr. 19 - 14600175 - 14674918 on NCBI Build GRCh38 | |
| Downstream | zinc finger protein 333 | ZNF333 | 67.4kb | Sense | Chr. 19 - 14689787 - 14733746 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SNORA104
Genomic Map