Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | elaC ribonuclease Z 1 | ELAC1 | D29 | -40.3kb | Sense | Chr. 18 - 50968017 - 50988120 on NCBI Build GRCh38 |
| Downstream | SMAD family member 4 | SMAD4 |
DPC4, JIP, MADH4, MYHRS |
1.5kb | Sense | Chr. 18 - 51030213 - 51085042 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map