Assay Details
| Gene Symbol: ATOH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006712122.3 | XP_006712185.1 | 2 - 3 | 1358 | 106 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
formation of primary germ layer
negative regulation of endothelial cell proliferation
transcription, DNA-templated
nervous system development
positive regulation of endothelial cell migration
cell differentiation
tube formation
positive regulation of endothelial cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
myoblast proliferation
SMAD protein signal transduction
transcription factor activity, sequence-specific DNA binding
activating transcription factor binding
protein dimerization activity
E-box binding
formation of primary germ layer
negative regulation of endothelial cell proliferation
transcription, DNA-templated
nervous system development
positive regulation of endothelial cell migration
cell differentiation
tube formation
positive regulation of endothelial cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
myoblast proliferation
SMAD protein signal transduction
transcription factor activity, sequence-specific DNA binding
activating transcription factor binding
protein dimerization activity
E-box binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000463422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000463422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000463422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000469442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000469442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000469442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000473116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000473116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000473116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000473116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000473116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000489682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | ENST00000489682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000306279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000463422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000463422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000463422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000469442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000469442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000469442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000473116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000473116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000473116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000473116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000473116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000489682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | ENST00000489682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | NM_032827.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008270.hg.1 | NM_032827.6 | Refseq |

ATOH8
Genomic Map