Assay Details
| Gene Symbol: ELFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052906.4 | NP_443138.2 | 2 - 3 | 326 | 93 | |
| GenBank mRNA | DA398675.1 | - | 2 - 3 | 327 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_052906.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |

ELFN2
Genomic Map