| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008941.hg.1 |
ENST00000341255.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008941.hg.1 |
ENST00000341255.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008941.hg.1 |
ENST00000341255.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008941.hg.1 |
ENST00000341255.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008941.hg.1 |
ENST00000341255.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008941.hg.1 |
NM_032287.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008941.hg.1 |
NM_032287.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008941.hg.1 |
NM_032287.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008941.hg.1 |
NM_032287.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008941.hg.1 |
ENST00000341255.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008941.hg.1 |
ENST00000341255.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008941.hg.1 |
ENST00000341255.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008941.hg.1 |
ENST00000341255.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008941.hg.1 |
ENST00000341255.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008941.hg.1 |
NM_032287.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008941.hg.1 |
NM_032287.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008941.hg.1 |
NM_032287.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008941.hg.1 |
NM_032287.2 |
Refseq |