Assay Details
| Gene Symbol: CMTM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181521.2 | NP_852662.1 | 4 | 4884 | 95 | |
| XM_011522882.1 | XP_011521184.1 | 4 | 4792 | 95 | ||
| GenBank mRNA | AK126493.1 | - | 1 | 2624 | 95 | |
| CR933620.1 | - | 1 | 3793 | 95 | ||
More Information
Set Membership: |
|

CMTM4
Genomic Map