Assay Details
| Gene Symbol: UBFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019116.2 | NP_061989.2 | 7 | 2494 | 66 | |
| XM_011545894.2 | XP_011544196.1 | 6 | 2417 | 66 | ||
| GenBank mRNA | AK025656.1 | - | 1 | 869 | 66 | |
| AK096189.1 | - | 3 | 1574 | 66 | ||
| AK124139.1 | - | 8 | 2270 | 66 | ||
More Information
Set Membership: |
|

UBFD1
Genomic Map