Assay Details
| Gene Symbol: PSME4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014614.2 | NP_055429.2 | 39 - 40 | 4580 | 89 | |
| XM_006711969.1 | XP_006712032.1 | 38 - 39 | 4157 | 89 | ||
| XM_011532705.2 | XP_011531007.1 | 40 - 41 | 4604 | 89 | ||
| XM_011532706.2 | XP_011531008.1 | 40 - 41 | 4604 | 89 | ||
| XM_011532707.2 | XP_011531009.1 | 39 - 40 | 4538 | 89 | ||
| GenBank mRNA | AK025517.1 | - | 8 - 9 | 924 | 89 | |
| AK026085.1 | - | 13 - 14 | 1577 | 89 | ||
| AY894754.1 | - | 38 - 39 | 4429 | 89 | ||
| AY894755.1 | - | 20 - 21 | 2268 | 89 | ||
| BC043602.1 | - | 4 - 5 | 374 | 89 | ||
| BC112169.1 | - | 38 - 39 | 4216 | 8327637 | 89 | |
| BC113668.1 | - | 38 - 39 | 4216 | 8322720 | 89 | |
| BC143739.1 | - | 38 - 39 | 4216 | 89 | ||
| D38521.1 | - | 39 - 40 | 4390 | 89 | ||
More Information
Set Membership: |
|

PSME4
Genomic Map