Assay Details
| Gene Symbol: ATP5G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125917.1 | - | 1 | 94 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010032.hg.1 | ENST00000416186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009350.hg.1 | ENST00000489560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009856.hg.1 | ENST00000567857.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000363251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000472782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000472782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000472782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000472782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000472782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009856.hg.1 | ENST00000567857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009350.hg.1 | ENST00000489560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010032.hg.1 | ENST00000416186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000284727.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000363251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000392541.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000409194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000472782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000472782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000472782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000472782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000472782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | ENST00000497075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001002258.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001190329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015012.hg.1 | NM_001689.4 | Refseq |

ATP5G3
Genomic Map