Assay Details
| Gene Symbol: HSPE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002157.2 | NP_002148.1 | 2 - 3 | 606 | 123 | |
| GenBank mRNA | AK312104.1 | - | 2 - 3 | 202 | 123 | |
| BC023518.2 | - | 2 - 3 | 280 | 4304224 | 123 | |
| BG188453.1 | - | 1 - 2 | 167 | 123 | ||
| CN264788.1 | - | 2 - 3 | 611 | 123 | ||
| CR407688.1 | - | 1 - 2 | 166 | 123 | ||
| U07550.1 | - | 2 - 3 | 207 | 123 | ||
| X75821.1 | - | 1 - 2 | 166 | 123 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
protein folding
activation of cysteine-type endopeptidase activity involved in apoptotic process
response to unfolded protein
chaperone mediated protein folding requiring cofactor
protein binding
ATP binding
poly(A) RNA binding
metal ion binding
unfolded protein binding
chaperone binding
osteoblast differentiation
protein folding
activation of cysteine-type endopeptidase activity involved in apoptotic process
response to unfolded protein
chaperone mediated protein folding requiring cofactor
protein binding
ATP binding
poly(A) RNA binding
metal ion binding
unfolded protein binding
chaperone binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011226.hg.1 | ENST00000443139.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011226.hg.1 | ENST00000443139.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008806.hg.1 | ENST00000455484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008806.hg.1 | ENST00000455484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008806.hg.1 | ENST00000455484.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008806.hg.1 | ENST00000455484.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011226.hg.1 | ENST00000443139.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011226.hg.1 | ENST00000443139.2 | Ensembl |

HSPE1
Genomic Map