Assay Details
| Gene Symbol: HOXD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK309681.1 | - | 1 - 2 | 105 | 107 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000498438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000498438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000498438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000498438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000498438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000619244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000619244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000619244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | ENST00000619244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | NM_021192.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | NM_021192.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | NM_021192.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | NM_021192.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | NM_021192.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016566.hg.1 | NM_021192.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000249504.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000498438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000498438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000498438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000498438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000498438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000619244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000619244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000619244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | ENST00000619244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | NM_021192.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | NM_021192.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | NM_021192.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | NM_021192.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | NM_021192.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016566.hg.1 | NM_021192.2 | Refseq |

HOXD11
Genomic Map