Assay Details
| Gene Symbol: TMA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF077202.1 | - | 2 - 3 | 104 | 91 | |
| AF161448.1 | - | 1 - 2 | 53 | 91 | ||
| AK126594.1 | - | 2 - 3 | 66 | 91 | ||
| AY508979.1 | - | 1 | 87 | 91 | ||
| AY794023.1 | - | 2 - 3 | 88 | 91 | ||
| BC001102.2 | - | 1 | 76 | 91 | ||
| BC080531.1 | - | 2 - 3 | 89 | 4875991 | 91 | |
| BU536461.1 | - | 2 - 3 | 133 | 91 | ||
More Information
Set Membership: |
|

TMA7
Genomic Map