Assay Details
| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX648832.1 | - | 1 - 2 | 49 | 79 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000492481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000492481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | ENST00000492481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000168148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000373368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000425558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000492481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000492481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | ENST00000492481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011148.hg.1 | NM_006944.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |

TMEM45A
Genomic Map