Assay Details
| Gene Symbol: SAMMSON | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110000.1 | 2 - 3 | 364 | 107 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000483525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | ENST00000488861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | NR_110000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007751.hg.1 | NR_110000.1 | Refseq |

SAMMSON
Genomic Map