| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000497374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000497374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000497374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000617024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000617024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
ENST00000620149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000359175.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000497137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000497374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000497374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000497374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000617024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000617024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
ENST00000620149.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012980.hg.1 |
NM_001040100.1 |
Refseq |