Assay Details
| Gene Symbol: FILIP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC020941.1 | - | 4 - 5 | 894 | 113 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000331335.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000331335.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000331335.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000331335.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000331335.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000331335.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000331335.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000331335.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000331335.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000331335.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000331335.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000354552.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000383694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000383694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000383694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000383694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000383694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000383694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000383694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000383694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000383694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000383694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000398326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000398326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000398326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000398326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000398326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000398326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000468533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000468533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000468533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000471562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000471562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000471562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000471562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000471562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000471562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000471562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000476723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000476723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000476723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000476723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000476723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000477258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000477258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000477258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000487087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000487087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000487087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000487087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000487087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000495625.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000495625.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000495625.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000495625.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000495625.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000495625.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | ENST00000495625.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001042459.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001042459.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001042459.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001042459.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001042459.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001042459.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001042459.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001042459.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001042459.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001042459.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001042459.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_001282794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_014890.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_014890.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_014890.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_014890.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_014890.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_014890.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_014890.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_014890.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_014890.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_014890.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011834.hg.1 | NM_182909.3 | Refseq |

FILIP1L
Genomic Map