Assay Details
| Gene Symbol: GLB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317040.1 | NP_001303969.1 | 2 - 3 | 363 | 110 | |
| GenBank mRNA | AK300753.1 | - | 2 - 3 | 279 | 110 | |
More Information
Set Membership: |
|

GLB1
Genomic Map