| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000456552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000456552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000456552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000456552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000456552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000479491.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000479491.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000479491.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000479491.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000495042.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000495042.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000495042.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000495042.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
ENST00000495042.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000442080.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000456552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000456552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000456552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000456552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000456552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000479491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000479491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000479491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000479491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000495042.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000495042.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000495042.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000495042.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
ENST00000495042.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013528.hg.1 |
NM_001146686.2 |
Refseq |