Assay Details
| Gene Symbol: NAALADL2-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046390.1 | 2 - 3 | 105 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAALADL2-AS3| Gene Symbol: NAALADL2-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046390.1 | 2 - 3 | 105 | 87 | ||
Set Membership: |
|