| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000526203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000526203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000526203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000526203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |