Assay Details
| Gene Symbol: LRRN4CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203422.3 | NP_981967.1 | 2 | 893 | 99 | |
| GenBank mRNA | AK291334.1 | - | 2 | 524 | 99 | |
| AL834319.1 | - | 2 | 722 | 99 | ||
| AY358354.1 | - | 2 | 566 | 99 | ||
| BC053902.1 | - | 2 | 596 | 6165542 | 99 | |
More Information
Set Membership: |
|

LRRN4CL
Genomic Map