Assay Details
| Gene Symbol: NUDT16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171905.1 | NP_001165376.1 | 4 | 5797 | 99 | |
| NM_152395.2 | NP_689608.2 | 3 | 5971 | 99 | ||
| NR_033268.1 | 2 | 6058 | 99 | |||
| GenBank mRNA | AK093567.1 | - | 1 | 1522 | 99 | |
More Information
Set Membership: |
|

NUDT16
Genomic Map