| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |