Assay Details
| Gene Symbol: LOC101927305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AW510858.1 | - | 1 - 2 | 308 | 83 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100006517.hg.1 | ENST00000623559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006517.hg.1 | ENST00000623559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006517.hg.1 | ENST00000623559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006517.hg.1 | ENST00000623559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006517.hg.1 | ENST00000624226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006517.hg.1 | ENST00000624226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006517.hg.1 | ENST00000624226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010197.hg.1 | ENST00000438072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010197.hg.1 | ENST00000438072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010197.hg.1 | ENST00000438072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010197.hg.1 | ENST00000438072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010197.hg.1 | ENST00000448491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010197.hg.1 | ENST00000448491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010197.hg.1 | ENST00000448491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | ENST00000507332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | ENST00000507332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | ENST00000507933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | ENST00000507933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | ENST00000507933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | ENST00000514222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | ENST00000514222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | ENST00000514222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | ENST00000514222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | ENST00000514222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | NR_125883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | NR_125883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | NR_125883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | NR_125883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | NR_125883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | NR_125884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | NR_125884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | NR_125884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008713.hg.1 | NR_125884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | ENST00000507332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | ENST00000507332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | ENST00000507933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | ENST00000507933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | ENST00000507933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | ENST00000514222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | ENST00000514222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | ENST00000514222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | ENST00000514222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | ENST00000514222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | NR_125883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | NR_125883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | NR_125883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | NR_125883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | NR_125883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | NR_125884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | NR_125884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | NR_125884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008713.hg.1 | NR_125884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010197.hg.1 | ENST00000438072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010197.hg.1 | ENST00000438072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010197.hg.1 | ENST00000438072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010197.hg.1 | ENST00000438072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010197.hg.1 | ENST00000448491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010197.hg.1 | ENST00000448491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010197.hg.1 | ENST00000448491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006517.hg.1 | ENST00000623559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006517.hg.1 | ENST00000623559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006517.hg.1 | ENST00000623559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006517.hg.1 | ENST00000623559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006517.hg.1 | ENST00000624226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006517.hg.1 | ENST00000624226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006517.hg.1 | ENST00000624226.1 | Ensembl |

LOC101927305
Genomic Map