Assay Details
| Gene Symbol: FAM47E-STBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242939.1 | NP_001229868.1 | 6 - 7 | 1074 | 95 | |
| GenBank mRNA | AK092277.1 | - | 6 - 7 | 1053 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK092277.1 transcripts.
Set Membership: |
|

FAM47E-STBD1
Genomic Map