Assay Details
| Gene Symbol: FGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK311683.1 | - | 1 - 2 | 124 | 69 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000302068.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000425838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000473984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000497097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000498375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000502545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | ENST00000509493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_001184741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009064.hg.1 | NM_005141.4 | Refseq |

FGB
Genomic Map