| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |