| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000459473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
NR_003689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000459473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000508179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
ENST00000509788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
NR_003689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006712.hg.1 |
NR_045196.2 |
Refseq |