Assay Details
| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323018.1 | NP_001309947.1 | 1 - 2 | 334 | 92 | |
| NM_001323020.1 | NP_001309949.1 | 1 - 2 | 334 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001323018.1, NM_001323020.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000423328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000423328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000423328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000423328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |

SETD9
Genomic Map